Assay Details
| Gene Symbol: SLURP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204521.2 | NP_989852.1 | 2 - 3 | 123 | 93 | |
| GenBank mRNA | AJ416114.2 | - | 1 - 2 | 76 | 93 | |
| BX276293.2 | - | 1 - 2 | 152 | 93 | ||
More Information
Set Membership: |
|

SLURP1
Genomic Map