Assay Details
| Gene Symbol: NR5A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205078.1 | NP_990409.1 | 3 - 4 | 462 | 56 | |
| XM_015290243.2 | XP_015145729.1 | 4 - 5 | 611 | 56 | ||
| XM_025152759.1 | XP_025008527.1 | 4 - 5 | 416 | 56 | ||
| XM_025152760.1 | XP_025008528.1 | 4 - 5 | 416 | 56 | ||
| XM_025152761.1 | XP_025008529.1 | 3 - 4 | 473 | 56 | ||
| XM_025152762.1 | XP_025008530.1 | 3 - 4 | 1734 | 56 | ||
| GenBank mRNA | AB002403.1 | - | 3 - 4 | 462 | 56 | |
More Information
Set Membership: |
|

NR5A2
Genomic Map