Assay Details
| Gene Symbol: ADORA2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205087.1 | NP_990418.1 | 1 - 2 | 416 | 87 | |
| GenBank mRNA | Y12601.1 | - | 416 | 87 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ADORA2B| Gene Symbol: ADORA2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205087.1 | NP_990418.1 | 1 - 2 | 416 | 87 | |
| GenBank mRNA | Y12601.1 | - | 416 | 87 | ||
Set Membership: |
|