Assay Details
| Gene Symbol: PPP1CB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205122.1 | NP_990453.1 | 7 - 8 | 1054 | 98 | |
| GenBank mRNA | D37987.1 | - | 7 - 8 | 1054 | 98 | |
More Information
Set Membership: |
|

PPP1CB
Genomic Map