Assay Details
Gene Symbol: SLC16A8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205140.2 | NP_990471.2 | 3 - 4 | 818 | 83 | |
GenBank mRNA | U15685.1 | - | 3 - 4 | 818 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
