Assay Details
| Gene Symbol: SNAI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205142.1 | NP_990473.1 | 2 - 3 | 613 | 64 | |
| GenBank mRNA | Y09905.1 | - | 613 | 64 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SNAI1| Gene Symbol: SNAI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205142.1 | NP_990473.1 | 2 - 3 | 613 | 64 | |
| GenBank mRNA | Y09905.1 | - | 613 | 64 | ||
Set Membership: |
|