Assay Details
| Gene Symbol: S100A11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205166.2 | NP_990497.1 | 2 - 3 | 222 | 97 | |
| GenBank mRNA | U77733.1 | - | 2 - 3 | 224 | 97 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

S100A11| Gene Symbol: S100A11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205166.2 | NP_990497.1 | 2 - 3 | 222 | 97 | |
| GenBank mRNA | U77733.1 | - | 2 - 3 | 224 | 97 | |
Set Membership: |
|