Assay Details
| Gene Symbol: MMP16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205197.2 | NP_990528.2 | 3 - 4 | 592 | 75 | |
| XM_025147058.1 | XP_025002826.1 | 3 - 4 | 631 | 75 | ||
| GenBank mRNA | AB260848.1 | - | 3 - 4 | 75 | ||
| CR389196.1 | - | 3 - 4 | 694 | 75 | ||
| U66463.1 | - | 3 - 4 | 75 | |||
More Information
Set Membership: |
|

MMP16
Genomic Map