Assay Details
| Gene Symbol: SLC2A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205209.1 | NP_990540.1 | 2 - 3 | 168 | 69 | |
| GenBank mRNA | L07300.1 | - | 2 - 3 | 168 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC2A1| Gene Symbol: SLC2A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205209.1 | NP_990540.1 | 2 - 3 | 168 | 69 | |
| GenBank mRNA | L07300.1 | - | 2 - 3 | 168 | 69 | |
Set Membership: |
|