Assay Details
| Gene Symbol: HSPA9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006147.1 | NP_001006147.1 | 10 - 11 | 1545 | 91 | |
| GenBank mRNA | AJ719486.1 | - | 10 - 11 | 1545 | 91 | |
More Information
Set Membership: |
|

HSPA9
Genomic Map