Assay Details
| Gene Symbol: PPP2CA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006152.1 | NP_001006152.1 | 2 - 3 | 526 | 63 | |
| GenBank mRNA | AJ719537.1 | - | 2 - 3 | 526 | 63 | |
More Information
Set Membership: |
|

PPP2CA
Genomic Map