Assay Details
| Gene Symbol: UBE2L3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006180.1 | NP_001006180.1 | 2 - 3 | 134 | 69 | |
| GenBank mRNA | AJ720047.1 | - | 2 - 3 | 134 | 69 | |
More Information
Set Membership: |
|

UBE2L3
Genomic Map