Assay Details
Gene Symbol: TMED2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006186.1 | NP_001006186.1 | 1 - 2 | 214 | 120 | |
XM_025155450.1 | XP_025011218.1 | 1 - 2 | 222 | 120 | ||
GenBank mRNA | AJ720174.1 | - | 1 - 2 | 214 | 120 | |
CR385511.1 | - | 1 - 2 | 197 | 120 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
