Assay Details
Gene Symbol: WDR5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006198.1 | NP_001006198.1 | 13 - 14 | 1136 | 127 | |
GenBank mRNA | AJ720488.1 | - | 13 - 14 | 1136 | 127 | |
BX931957.2 | - | 13 - 14 | 1060 | 127 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
