Assay Details
Gene Symbol: WDR5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006198.1 | NP_001006198.1 | 5 - 6 | 590 | 96 | |
GenBank mRNA | AJ720488.1 | - | 5 - 6 | 590 | 96 | |
BX931957.2 | - | 5 - 6 | 514 | 96 | ||
BX935058.2 | - | 5 - 6 | 589 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() |
