Assay Details
Gene Symbol: WDR5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006198.1 | NP_001006198.1 | 8 - 9 | 819 | 73 | |
GenBank mRNA | AJ720488.1 | - | 8 - 9 | 819 | 73 | |
BX931957.2 | - | 8 - 9 | 743 | 73 | ||
BX935058.2 | - | 8 - 9 | 818 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
