Assay Details
| Gene Symbol: GOSR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006222.1 | NP_001006222.1 | 7 - 8 | 539 | 84 | |
| XM_015295771.2 | XP_015151257.1 | 7 - 8 | 572 | 84 | ||
| GenBank mRNA | AJ720233.1 | - | 7 - 8 | 539 | 84 | |
More Information
Set Membership: |
|

GOSR1
Genomic Map