Assay Details
Gene Symbol: TMPO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006235.1 | NP_001006235.1 | 3 - 4 | 625 | 116 | |
XM_015284619.2 | XP_015140105.1 | 3 - 4 | 669 | 116 | ||
XM_015284625.2 | XP_015140111.1 | 3 - 4 | 668 | 116 | ||
XM_015284631.2 | XP_015140117.1 | 3 - 4 | 671 | 116 | ||
XM_015284637.2 | XP_015140123.1 | 3 - 4 | 671 | 116 | ||
XM_015284646.2 | XP_015140132.1 | 3 - 4 | 666 | 116 | ||
XM_015284657.2 | XP_015140143.1 | 3 - 4 | 666 | 116 | ||
XM_015284666.2 | XP_015140152.1 | 3 - 4 | 629 | 116 | ||
GenBank mRNA | AJ719348.1 | - | 3 - 4 | 625 | 116 | |
AJ719530.1 | - | 3 - 4 | 625 | 116 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
