Assay Details
| Gene Symbol: TAF13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006252.1 | NP_001006252.1 | 2 - 3 | 121 | 72 | |
| GenBank mRNA | AJ719329.1 | - | 2 - 3 | 117 | 72 | |
| AJ719754.1 | - | 2 - 3 | 121 | 72 | ||
| CR354130.1 | - | 1 - 2 | 83 | 72 | ||
| CR388997.1 | - | 2 - 3 | 107 | 72 | ||
| CR389522.1 | - | 2 - 3 | 127 | 72 | ||
More Information
Set Membership: |
|

TAF13
Genomic Map