Assay Details
| Gene Symbol: PRPS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006264.1 | NP_001006264.1 | 6 - 7 | 771 | 65 | |
| XM_015273663.2 | XP_015129149.1 | 5 - 6 | 753 | 65 | ||
| XM_015273666.2 | XP_015129152.1 | 5 - 6 | 617 | 65 | ||
| GenBank mRNA | AJ720935.1 | - | 6 - 7 | 771 | 65 | |
| CR354205.1 | - | 5 - 6 | 733 | 65 | ||
More Information
Set Membership: |
|

PRPS2
Genomic Map