Assay Details
| Gene Symbol: CKAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006274.1 | NP_001006274.1 | 6 - 7 | 1423 | 65 | |
| GenBank mRNA | AJ720484.1 | - | 6 - 7 | 1423 | 65 | |
More Information
Set Membership: |
|

CKAP2
Genomic Map