Assay Details
| Gene Symbol: UQCC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006285.1 | NP_001006285.1 | 6 - 7 | 484 | 64 | |
| XM_015296246.2 | XP_015151732.1 | 6 - 7 | 586 | 64 | ||
| XM_025142189.1 | XP_024997957.1 | 6 - 7 | 772 | 64 | ||
| GenBank mRNA | AJ720641.1 | - | 6 - 7 | 484 | 64 | |
More Information
Set Membership: |
|

UQCC1
Genomic Map