Assay Details
| Gene Symbol: CHMP4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006286.1 | NP_001006286.1 | 3 - 4 | 674 | 71 | |
| GenBank mRNA | AJ721089.1 | - | 3 - 4 | 674 | 71 | |
More Information
Set Membership: |
|

CHMP4B
Genomic Map