Assay Details
Gene Symbol: STX16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006295.1 | NP_001006295.1 | 4 - 5 | 601 | 75 | |
XM_015296405.1 | XP_015151891.1 | 4 - 5 | 555 | 75 | ||
XM_015296406.1 | XP_015151892.1 | 3 - 4 | 544 | 75 | ||
GenBank mRNA | AJ720197.1 | - | 4 - 5 | 601 | 75 | |
CR386335.1 | - | 1 - 2 | 69 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
