Assay Details
| Gene Symbol: RTFDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006297.2 | NP_001006297.2 | 1 - 2 | 124 | 73 | |
| XM_015296411.2 | XP_015151897.1 | 1 - 2 | 116 | 73 | ||
| GenBank mRNA | AJ720403.1 | - | 1 - 2 | 127 | 73 | |
More Information
Set Membership: |
|

RTFDC1
Genomic Map