Assay Details
| Gene Symbol: ADPRHL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006312.1 | NP_001006312.1 | 2 - 3 | 331 | 66 | |
| XM_025142993.1 | XP_024998761.1 | 2 - 3 | 319 | 66 | ||
| GenBank mRNA | AJ720933.1 | - | 2 - 3 | 331 | 66 | |
| BX930057.1 | - | 2 - 3 | 324 | 66 | ||
More Information
Set Membership: |
|

ADPRHL2
Genomic Map