Assay Details
| Gene Symbol: CUTC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006503.1 | NP_001006503.1 | 5 - 6 | 436 | 52 | |
| XM_015288626.2 | XP_015144112.1 | 3 - 4 | 52 | |||
| XM_015288627.2 | XP_015144113.1 | 4 - 5 | 52 | |||
| XM_015288629.2 | XP_015144115.1 | 3 - 4 | 52 | |||
| XM_025151603.1 | XP_025007371.1 | 5 - 6 | 52 | |||
| GenBank mRNA | AJ720183.1 | - | 5 - 6 | 436 | 52 | |
| CR391564.1 | - | 1 - 2 | 114 | 52 | ||
More Information
Set Membership: |
|

CUTC
Genomic Map