Assay Details
| Gene Symbol: UBE2F | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006512.1 | NP_001006512.1 | 6 - 7 | 439 | 76 | |
| XM_015289164.2 | XP_015144650.1 | 6 - 7 | 493 | 76 | ||
| GenBank mRNA | AJ719958.1 | - | 6 - 7 | 439 | 76 | |
More Information
Set Membership: |
|

UBE2F
Genomic Map