Assay Details
Gene Symbol: DDX19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006568.1 | NP_001006568.1 | 1 - 2 | 83 | 68 | |
XM_015296933.2 | XP_015152419.1 | 1 - 2 | 155 | 68 | ||
GenBank mRNA | AJ719463.1 | - | 1 - 2 | 83 | 68 | |
AJ719483.1 | - | 1 - 2 | 131 | 68 | ||
BX936036.1 | - | 1 - 2 | 100 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
