Assay Details
| Gene Symbol: ACAA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006571.2 | NP_001006571.1 | 7 - 8 | 910 | 69 | |
| GenBank mRNA | AJ719616.1 | - | 7 - 8 | 912 | 69 | |
More Information
Set Membership: |
|

ACAA2
Genomic Map