Assay Details
Gene Symbol: NSA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006579.1 | NP_001006579.1 | 3 - 4 | 370 | 82 | |
GenBank mRNA | AJ719405.1 | - | 3 - 4 | 370 | 82 | |
CR388679.1 | - | 3 - 4 | 412 | 82 | ||
CR523694.1 | - | 4 - 5 | 283 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
