Assay Details
Gene Symbol: NSA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006579.1 | NP_001006579.1 | 5 - 6 | 738 | 107 | |
GenBank mRNA | AJ719405.1 | - | 5 - 6 | 738 | 107 | |
CR523694.1 | - | 6 - 7 | 651 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
