Assay Details
| Gene Symbol: RIOK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006581.2 | NP_001006581.2 | 7 - 8 | 1189 | 68 | |
| GenBank mRNA | AJ719586.1 | - | 7 - 8 | 932 | 68 | |
| AJ731428.1 | - | 1 - 2 | 80 | 68 | ||
More Information
Set Membership: |
|

RIOK2
Genomic Map