Assay Details
| Gene Symbol: AP3S1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006586.1 | NP_001006586.1 | 4 - 5 | 414 | 77 | |
| GenBank mRNA | AJ720492.1 | - | 4 - 5 | 414 | 77 | |
More Information
Set Membership: |
|

AP3S1
Genomic Map