Assay Details
| Gene Symbol: HACD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007828.1 | NP_001007829.1 | 2 - 3 | 183 | 74 | |
| GenBank mRNA | AJ719527.1 | - | 2 - 3 | 183 | 74 | |
More Information
Set Membership: |
|

HACD3
Genomic Map