Assay Details
| Gene Symbol: PAIP2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007832.1 | NP_001007833.1 | 3 - 4 | 454 | 74 | |
| XM_025154840.1 | XP_025010608.1 | 3 - 4 | 663 | 74 | ||
| GenBank mRNA | AJ720358.1 | - | 3 - 4 | 454 | 74 | |
More Information
Set Membership: |
|

PAIP2B
Genomic Map