Assay Details
| Gene Symbol: SNRPD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007838.1 | NP_001007839.1 | 3 - 4 | 400 | 58 | |
| XM_025155502.1 | XP_025011270.1 | 3 - 4 | 406 | 58 | ||
| GenBank mRNA | AJ719876.1 | - | 3 - 4 | 400 | 58 | |
| CR388618.1 | - | 3 - 4 | 382 | 58 | ||
More Information
Set Membership: |
|

SNRPD3
Genomic Map