Assay Details
| Gene Symbol: YAF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007850.1 | NP_001007851.1 | 2 - 3 | 298 | 69 | |
| GenBank mRNA | AJ720159.1 | - | 2 - 3 | 298 | 69 | |
More Information
Set Membership: |
|

YAF2
Genomic Map