Assay Details
| Gene Symbol: FGFR1OP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007855.1 | NP_001007856.1 | 4 - 5 | 459 | 87 | |
| XM_015290277.2 | XP_015145763.1 | 4 - 5 | 506 | 87 | ||
| GenBank mRNA | AJ720090.1 | - | 4 - 5 | 459 | 87 | |
More Information
Set Membership: |
|

FGFR1OP2
Genomic Map