Assay Details
| Gene Symbol: TCEB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007888.1 | NP_001007889.1 | 3 - 4 | 276 | 110 | |
| GenBank mRNA | AJ720347.1 | - | 3 - 4 | 276 | 110 | |
| CR390316.1 | - | 3 - 4 | 281 | 110 | ||
More Information
Set Membership: |
|

TCEB1
Genomic Map