Assay Details
| Gene Symbol: KCTD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007913.1 | NP_001007914.1 | 2 - 3 | 436 | 81 | |
| GenBank mRNA | AJ720572.1 | - | 2 - 3 | 436 | 81 | |
More Information
Set Membership: |
|

KCTD2
Genomic Map