Assay Details
Gene Symbol: CHP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007930.1 | NP_001007931.1 | 1 - 2 | 187 | 74 | |
GenBank mRNA | AJ719540.1 | - | 1 - 2 | 187 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
