Assay Details
| Gene Symbol: BRMS1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007935.1 | NP_001007936.1 | 1 - 2 | 201 | 87 | |
| GenBank mRNA | AJ719715.1 | - | 1 - 2 | 201 | 87 | |
More Information
Set Membership: |
|

BRMS1L
Genomic Map