Assay Details
| Gene Symbol: SLC25A15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008442.1 | NP_001008442.1 | 5 - 6 | 722 | 70 | |
| XM_015275733.2 | XP_015131219.1 | 5 - 6 | 701 | 70 | ||
| XM_015275743.2 | XP_015131229.1 | 5 - 6 | 778 | 70 | ||
| XM_015275752.2 | XP_015131238.1 | 5 - 6 | 924 | 70 | ||
| XM_015275754.2 | XP_015131240.1 | 4 - 5 | 653 | 70 | ||
| GenBank mRNA | AJ720668.1 | - | 5 - 6 | 722 | 70 | |
More Information
Set Membership: |
|

SLC25A15
Genomic Map