Assay Details
| Gene Symbol: LZIC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008450.1 | NP_001008450.1 | 6 - 7 | 585 | 74 | |
| XM_015296891.2 | XP_015152377.1 | 6 - 7 | 781 | 74 | ||
| XM_015296892.2 | XP_015152378.1 | 6 - 7 | 711 | 74 | ||
| XM_025142520.1 | XP_024998288.1 | 6 - 7 | 618 | 74 | ||
| XM_025142521.1 | XP_024998289.1 | 6 - 7 | 758 | 74 | ||
| GenBank mRNA | AJ719972.1 | - | 6 - 7 | 585 | 74 | |
More Information
Set Membership: |
|

LZIC
Genomic Map