Assay Details
| Gene Symbol: TRAPPC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008451.1 | NP_001008451.1 | 4 - 5 | 485 | 117 | |
| GenBank mRNA | AJ720927.1 | - | 4 - 5 | 485 | 117 | |
More Information
Set Membership: |
|

TRAPPC3
Genomic Map