Assay Details
| Gene Symbol: CCND3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008453.1 | NP_001008453.1 | 689 | 74 | ||
| XM_015298952.2 | XP_015154438.1 | 526 | 74 | |||
| XM_015298953.2 | XP_015154439.1 | 525 | 74 | |||
| GenBank mRNA | AJ720095.1 | - | 689 | 74 | ||
More Information
Set Membership: |
|

CCND3
Genomic Map