Assay Details
| Gene Symbol: ATP6V0D2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008455.1 | NP_001008455.1 | 7 - 8 | 964 | 79 | |
| GenBank mRNA | AJ721124.1 | - | 7 - 8 | 964 | 79 | |
More Information
Set Membership: |
|

ATP6V0D2
Genomic Map