Assay Details
| Gene Symbol: PPIL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008475.1 | NP_001008475.1 | 6 - 7 | 507 | 68 | |
| GenBank mRNA | AJ719632.1 | - | 6 - 7 | 507 | 68 | |
| CR387884.1 | - | 5 - 6 | 576 | 68 | ||
| CR389824.1 | - | 6 - 7 | 491 | 68 | ||
More Information
Set Membership: |
|

PPIL3
Genomic Map