Assay Details
Gene Symbol: DR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008478.1 | NP_001008478.1 | 2 - 3 | 783 | 71 | |
GenBank mRNA | AJ719281.1 | - | 2 - 3 | 783 | 71 | |
BX931827.2 | - | 2 - 3 | 478 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
